Learn More
Statebourne Cryogenics™ Cryolab™ Series Cryogenic Dewars
Excellent for the storage and handling of liquid nitrogen in the laboratory. Statebourne Cryogenics™ Cryolab™ Series Cryogenic Dewars have an all-aluminum construction.
en la oferta
Especificaciones
Incluye Neckplugs and dipstick
Para utilizar con (aplicación) Storage of liquid nitrogen
Material Aluminum
Ver más especificaciones
Descripción
Descripción
  • All units guaranteed for three years
  • ISO 9001 designed and manufactured
  • Supplied with neckplugs and dipstick
  • Comprehensive range of accessories available
Especificaciones
Especificaciones
Neckplugs and dipstick
Storage of liquid nitrogen
Aluminum