missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRYBA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15750-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
the domit structural components of the vertebrate eye lens.
Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of the vertebrate eye, which function to maintain the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also defined as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Beta-crystallins, the most heterogeneous, differ by the presence of the C-terminal extension (present in the basic group but absent in the acidic group). Beta-crystallins form aggregates of different sizes and are able to form homodimers through self-association or heterodimers with other beta-crystallins. This gene is a beta acidic group member. Three alternatively spliced transcript variants encoding identical proteins have been reported.Especificaciones
CRYBA2 | |
Polyclonal | |
Unconjugated | |
CRYBA2 | |
Beta A2 crystallin, Beta crystallin A2, CRYBA2, crystallin, beta A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12958, 1412, 286925 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P53672, Q9JJV1 | |
CRYBA2 | |
CRYBA2 Fusion Protein Ag8222 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |