missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRP Mouse anti-Human, Mouse, Porcine, Rat, Clone: 3F10A11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66250-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
CRP (C-reactive protein) is a major cyclic, pentameric acute phase protein compound consisting of five identical, noncovalently bound, nonglycosylated subunits. Eeach subunit is made up of 206 amino acids, and has a molecular weight of 24 kDa. CRP is produced by the liver, and its plasma levels rise dramatically during inflammatory processes occurring in the body. CRP is an initiator of classical complement cascade, binds to several nuclear components (chromatin, histones, etc.), plays an important role in innate immunity, and mediates activities associated with pre-immune nonspecific host resistance. Specifically, there is a 100-1000 fold increase of CRP in response to injury, infection or inflammation. CRP is opsonic, an initiator of the classical complement cascade, and an activator of monocytes/macrophages. CRP also binds to several nuclear components including chromatin, histones and snRNP, suggesting that it may play a role as a scavenger during cell necrosis. CRP is involved in several host defense related functions based on its ability to recognize foreign pathogens and damaged cells of the host, and initiates pathogen elimination by interacting with humoral and cellular effector systems in the blood. Patients with elevated basal levels of CRP have been shown to be an increased risk for hypertension and cardiovascular disease.Especificaciones
CRP | |
Monoclonal | |
1.6 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O19062, P02741, P14847, P48199 | |
Crp | |
CRP Fusion Protein Ag9883 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
3F10A11 | |
Unconjugated | |
CRP | |
C reactive protein, CRP, PTX1 | |
Mouse | |
Protein A | |
RUO | |
100620468, 12944, 1401, 25419 | |
-20°C | |
Liquid |