missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRMP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18969-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
DPYSL3 is necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. DPYSL3 plays a role in axon guidance, neuronal growth cone collapse and cell migration.Especificaciones
CRMP4 | |
Polyclonal | |
Unconjugated | |
DPYSL3 | |
CRMP 4, CRMP4, dihydropyrimidinase like 3, DPYSL3, DRP 3, DRP3, LCRMP, ULIP, ULIP 1, ULIP1, Unc 33 like phosphoprotein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1809, 22240, 25418 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14195, Q62188, Q62952 | |
DPYSL3 | |
CRMP4 Fusion Protein Ag5299 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |