Learn More
Code Corporation™ CR2700 Handheld Wireless Handle Barcode Scanner with Charging Station Bluetooth
Product Code. 17953862 Shop All Brady Products
Change view
Click to view available options
Connection Type:
Bluetooth
Unit Size:
Each
Product Code. Connection Type Unit Size
17953862 Bluetooth Each
1 options
This item is not returnable. View return policy
Product Code. 17953862 Supplier Code Corporation™ Supplier No. 176516
  • ONLINE EXCLUSIVE PRICE
    1555.00€
    1134.00€ / paquete de 1
    Ahorro 421.00€
    27% Descuento
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Easily read wide and small barcodes with the ultra-fast CR2700 Handled Barcode Scanner. This aggressive, omnidirectional scanner eliminates repositioning and time-consuming rescans of barcodes. Kit features a Charging Station.
  • Use this comprehensive kit to get your scanning processes started
  • Includes everything you need to set up a scanning workflow
  • Uses B27 batteries with built-in power gauge
  • IP65 seal rating against dust and liquid ingress
  • CodeShield™ housing for disinfectant and chemical wipedown
  • Unequaled performance reading on curved and shiny surfaces
  • USB, Bluetooth™5 Low Energy (for enhanced data security, extended battery life and less downtime)
  • Customizable visual, audible and haptic feedback with multiple programmable buttons
  • Trace any troubles: Trackable service dates and battery statistics to help IT deploy and manage their scanner fleet
Specifications
Height (Metric) 135 mm
Height (English) 5.3 in.
Width (Metric) 53.34 mm
Width (English) 2.1 in.
Depth (Metric) 131 mm
Depth (English) 5.2 in.
Weight (Metric) 177 g
Weight (English) 0.29 lb.
Field View High Density Field: 30° Horizontal by 20° Vertical, Wide Field: 50° Horizontal by 33.5° Vertical
Includes CR2700 Scanner Handled Version, B27 Battery, Inductive Charging Station with Desktop Mount, 6 ft. USB Charging Cable
Type Barcode Scanner
Connection Type Bluetooth
Data Memory 1 MB NOR Flash, 16 MB RAM
Power Supply Rechargeable Battery
Resolution High Density Field: 960 x 640; Wide Field: 960 x 640
Alternate Part Number CR2702-200-A272-C36-MB6-C286