missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPZ Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15944-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
CPZ is a member of the metallocarboxypeptidase family. This enzyme displays carboxypeptidase activity towards substrates with basic C-terminal residues. It is most active at neutral pH and is inhibited by active site-directed inhibitors of metallocarboxypeptidases.Especificaciones
CPZ | |
Polyclonal | |
Unconjugated | |
CPZ | |
carboxypeptidase Z, CPZ | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
242939, 83575, 8532 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O54858, Q66K79, Q8R4V4 | |
CPZ | |
CPZ Fusion Protein Ag8429 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |