missing translation for 'onlineSavingsMsg'
Learn More

CPT1B Antibody (43), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90486B

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215829

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

CPT1B Monoclonal specifically detects CPT1B in Mouse samples. It is validated for ELISA.
Especificaciones

Especificaciones

CPT1B
Monoclonal
Biotin
carnitine O-palmitoyltransferase 1, muscle isoform, carnitine O-palmitoyltransferase I, mitochondrial muscle isoform, Carnitine O-palmitoyltransferase I, muscle isoform, Carnitine palmitoyltransferase 1B, carnitine palmitoyltransferase 1B (muscle), Carnitine palmitoyltransferase I-like protein, CPT I, CPT1M, CPT1-MMCCPT1, CPTI, CPTI-M, EC 2.3.1, EC 2.3.1.21, FLJ55729, FLJ58750, KIAA1670, MCPT1, M-CPT1
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CPT1B (NP_032596.1; Met 1-Lys 246).
0.1 mL
Lipid and Metabolism
1375
Store at 4°C in the dark.
IgG
ELISA
43
PBS
Rabbit
Protein A purified
RUO
Primary
Mouse
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales