missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPNE9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20098-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is specific to CPNE9.
The exact functions of CPNE9 remain unknown.Especificaciones
CPNE9 | |
Polyclonal | |
Unconjugated | |
CPNE9 | |
Copine 9, copine family member IX, Copine IX, CPNE9, KIAA4217 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
151835, 211232, 297516 | |
-20°C | |
Liquid |
Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q1RLL3, Q5BJS7, Q8IYJ1 | |
Cpne9 | |
Peptide | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |