missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ CPC™ Quick Disconnect Valved Polypropylene Insert

Quick Disconnect couplings feature a thumb latch for easy assembly

Marca:  Cole-Parmer™ 31303-86

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15457093

  • 17.51€ / cada uno


Consúltenos : 902 239 303 
Añadir a la cesta

Descripción

Descripción

  • High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately)
  • Plug insert into coupling body to connect
  • Fitting halves lock into place immediately
  • To disconnect fitting halves, simply press the button on the coupling body
  • Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged
  • All fitting bodies have 316 SS latch springs and EPDM seals
Especificaciones

Especificaciones

3/8 in. Quick-Disconnect
1/2 in.
Valved Polypropylene Insert
60 psi
Gamma or e-beam
Polypropylene, EPDM
71°C
Videos