Learn More
Cole-Parmer™ CPC Miniature Quick-Disconnect Valved Male Hose Barb
Quick Disconnect couplings feature a thumb latch for easy assembly.
Marca:  Cole-Parmer™ 06363-83
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15456523
  • 32.42€ / paquete de 10
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • CPC Miniature Quick-Disconnect Valved Male Hose Barb
  • Standard flow quick-disconnect couplings require a body and an insert of the same flow size
  • Plug insert into body to connect fitting
  • Fitting halves lock into place immediately
  • To disconnect fitting halves, simply press the thumb latch on the coupling body
Especificaciones
Especificaciones
Quick-Disconnect x Barb Connection
1.905 cm
1/16 in.
1/16 in.
Polycarbonate
Valved Male Hose Barb
3/32 in.