Learn More
Cole-Parmer™ CPC™ High-Flow Quick-Disconnect Straight-through Hose Barb Fittings
Designed to enable enhanced flow in a coupling with a 3/8 inch flow path, the HFC series is available in polypropylene and polysulfone for a wide variety of applications
Marca:  Cole-Parmer™ 31307-42
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15497133
Este producto se ha descatalogado y ya no está en venta. Consulte el producto para saber si se ofrecen alternativas o póngase en contacto con nuestro equipo de Atención al Cliente en el número 902 239 303.
Descripción
Descripción
  • High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately).
  • Plug insert into coupling body to connect.
  • Fitting halves lock into place immediately.
  • To disconnect fitting halves, simply press the button on the coupling body.
  • HFC series has an ergonomic, easy-to-operate design and comes standard with a shrouded latch to protect against accidental disconnect.
  • UV-resistant polysulfone.
  • Couplings of UV-resistant polysulfone and with BSP threaded terminations are also available; contact Cole-Parmer for more information.
Especificaciones
Especificaciones
Gray
Autoclavable
Valved Bulkhead Panel Mount Hose Barb Body
137°C
-40°C
27.5 in. Hg
Autoclave, Ethylene Oxide or Gamma
Quick-Disconnect x Barb Connection
Polysulfone
125 psi
-40°F
5/8 in.
5/8 in.
3/8 in. Quick Disconnect