Learn More
Cole-Parmer™ CPC™ High-Flow Quick-Disconnect Straight-through Elbow Fittings
Designed to enable enhanced flow in a coupling with a 3/8 inch flow path, the HFC series is available in polypropylene and polysulfone for a wide variety of applications
10.69€
Especificaciones
Tipo Valved Elbow Fittings
Material Polysulfone
Esterilizable en autoclave Yes
Color Grey
Método de esterilización Autoclave, Ethylene Oxide (ETO) or Gamma
Ver más especificaciones
Productos 1
Código de producto Marca Tamaño de los tubos Precio Cantidad y disponibilidad  
Código de producto Marca Tamaño de los tubos Precio Cantidad y disponibilidad  
15447143 Cole-Parmer™
31307-51
1/2 in. 10.69€
cada uno Ver productos alternativos
Ahorre hasta 
»
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately).
  • Plug insert into coupling body to connect.
  • Fitting halves lock into place immediately.
  • To disconnect fitting halves, simply press the button on the coupling body.
  • HFC series has an ergonomic, easy-to-operate design and comes standard with a shrouded latch to protect against accidental disconnect.
  • UV-resistant polysulfone.
  • Couplings of UV-resistant polysulfone and with BSP threaded terminations are also available; contact Cole-Parmer for more information.
Especificaciones
Especificaciones
Valved Elbow Fittings
Yes
Autoclave, Ethylene Oxide (ETO) or Gamma
137°C
Polysulfone
Grey
-40°C