Learn More
Cole-Parmer™ CPC™ High-Flow Quick-Disconnect Fitting with Bulkhead Panel Mount Hose Barb Bodies
Eliminate leaks with these valved couplings
Marca:  Cole-Parmer™ 31307-25
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15497123
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Descripción
Descripción
  • High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately).
  • Plug insert into coupling body to connect.
  • Fitting halves lock into place immediately.
  • To disconnect fitting halves, simply press the button on the coupling body.
  • Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged.
  • All fitting bodies have 316 SS latch springs and EPDM seals.
Especificaciones
Especificaciones
Gray
Autoclavable
Valved Bulkhead Panel Mount Hose Barb Body
137°C
-40°C
27.5 in. Hg
3/8 in. Quick Disconnect
Quick-Disconnect x Barb Connection
Polysulfone
125 psi
-40°F
3/8 in.
3/8 in.
Autoclave, Ethylene Oxide or Gamma