Learn More
3M™ Coupling Plugs
Product Code. 12374679 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
1/4 in. BSP CEJN '342' regulators
1/4 in. BSP Rectus 25 regulators
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12374679 1/4 in. BSP Rectus 25 regulators Each
12364679 1/4 in. BSP CEJN '342' regulators Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12374679 Supplier 3M™ Supplier No. 7000044241
  • ONLINE EXCLUSIVE PRICE
    99.40€
    23.47€ / paquete de 1
    Ahorro 75.93€
    76% Descuento
Fecha estimada de envoi: 18-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Product Type Coupling
For Use With (Application) Connect compressed air supply tubes to air regulators
For Use With (Equipment) 1/4 in. BSP Rectus 25 regulators