Learn More
Corning™ SF Medium
Based on a proprietary combination of classical media formulations
Marca:  Corning™ 40-102-CV
Detalles adicionales : Peso : 3.76000kg
 Ver más versiones de este producto
Código de producto. 15333701
  • 157.15€ / paquete de 6
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Contains a mixture of select trace elements and high molecular carbohydrates, extra vitamins, animal origin-free protein, and a small amount of high quality bovine serum albumin (1.0 g/L)
  • Supports superior growth and viability over long-term passages in hybridomas, as well as suspension and adherent cell cultures
  • Formulation contains L-glutamine
Offers a viable alternative to FBS for many hybridomas and works on a broad range of suspension and attachment cells; can be cell line dependent
Especificaciones
Especificaciones
Sterile
SF Medium, With L-Glutamine; without Phenol Red
2° to 8°C
7.2 to 7.6
6 x 500mL
Phenol Red
Videos