Learn More
Corning™ L-glutamine Powder
An essential amino acid and a key component of culture media
Marca:  Corning™ 61-030-RO
Detalles adicionales : Peso : 0.54000kg
 Ver más versiones de este producto
Código de producto. 15333751
  • 266.20€ / 500 g
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
  • Serves as major energy source for propagating cells
  • Molecular Weight: 146.15
Especificaciones
Especificaciones
Powder
5 to 6
L-Glutamine
Plastic Bottle
500g
2° to 8°C
Videos