missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihidróxido de carbonato de cobre(II), polvo verde, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Cantidad:
500 g
2 kg
Description
Copper(II) carbonate dihydroxide is used as pigments and in treatment for copper deficiency in ruminants. It is also used in pyrotechnics, in sweetening of petrol sour crude stock and animal and poultry feeds. It finds application in some types of make-up such as lipstick. In analytical chemistry, it is used as a reagent for analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nombre del producto químico o material | Copper(II) carbonate dihydroxide |
Punto de fusión | 200°C (decomposition) |
Nombre de nota | Cu 55% min. |
Color | Verde |
Forma física | Polvo |
Cantidad | 2 kg |
Información de solubilidad | Soluble in acid,ammonia,ammonium salt and alkali metal carbonate solution. Insoluble in water and alcohol. |
Formula Weight (peso de la fórmula) | 221.11 |
Densidad | 4 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'