Learn More
Copper(I) chloride, 99.999% (metals basis), Alfa Aesar™
Marca: Alfa Aesar 014644.22
Detalles adicionales : CAS : 7758-89-6 Peso : 0.10000kg Transport : UN number : 2802 Chem class : 8 Pack group : III
Cantidad. | 100g |
---|
Identificadores de productos químicos
7758-89-6 | |
98.996 | |
OXBLHERUFWYNTN-UHFFFAOYSA-M | |
62652 | |
chlorocopper |
ClCu | |
MFCD00010971 | |
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, copper i chloride, chlorid medny czech, copper i chloride | |
CHEBI:53472 | |
Cl[Cu] |
Especificación
Copper(I) chloride | |
99.999% (Metal basis) | |
MFCD00010971 | |
OXBLHERUFWYNTN-UHFFFAOYSA-M | |
chlorocopper | |
62652 | |
98.99 | |
100g |
7758-89-6 | |
ClCu | |
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, copper i chloride, chlorid medny czech, copper i chloride | |
Cl[Cu] | |
98.996 | |
CHEBI:53472 | |
Powder |