Learn More
Conventional Closure Polyethylene Terephthalate Blood Tube
Product Code. 10184469
Change view
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Pack of 100
Product Code. Capacity (Metric) Unit Size
10184469 10 mL Pack of 100
1 options
This item is not returnable. View return policy
Product Code. 10184469 Supplier BD Vacutainer Supplier No. 367526
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Specifications
Product Type Blood Collection Tube
Additive Lithium Heparin
Material PET
Closure Type Traditional Cap
Capacity (Metric) 10 mL
Closure Color Green
Label Type Paper
Diameter (Metric) Outer 16 mm