missing translation for 'onlineSavingsMsg'
Learn More

Complexin-2 Antibody (021), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90019JF549

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215968

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Complexin-2 Monoclonal specifically detects Complexin-2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

Complexin-2
Monoclonal
Janelia Fluor 549
50mM Sodium Borate
Rabbit
Protein A purified
RUO
10814
Store at 4°C in the dark.
IgG
Flow Cytometry, Immunofluorescence
21
Flow Cytometry, Immunocytochemistry/Immunofluorescence
complexin 2, Complexin II, complexin-2, CPX II, CPX2, CPX-2, DKFZp547D155, Hfb1, MGC138492, synaphin 1,921-L, synaphin-1
This antibody was obtained from a rabbit immunized with purified, recombinant Human Complexin-2 (Q6PUV4-1; Asp2-Lys134).
0.1 mL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales