missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C5aR1 Antibody (P12/1), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NB100-66539FR

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215480

  • 566.94€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Complement Component C5aR1 Monoclonal specifically detects Complement Component C5aR1 in Human, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones

Especificaciones

Complement Component C5aR1
Monoclonal
DyLight 680
P21730
C5AR1
C5aR - peptide: Met1 - Asn31
0.1 mL
GPCR, Immunology, Innate Immunity
728
Human, Rhesus Macaque
Purified
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
P12/1
Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
C5A, C5a anaphylatoxin chemotactic receptor, C5aR, C5a-R, C5ARC5a anaphylatoxin receptor, C5R1C5a ligand, CD88, CD88 antigen, complement component 5 receptor 1, complement component 5 receptor 1 (C5a ligand), complement component 5a receptor 1
Mouse
Protein G purified
RUO
Primary
Recognizes the C5a receptor (C5aR) CD88, which is predominantly expressed on cells of the myeloid lineage. Clone P12/1 was raised against a synthetic peptide comprising the N-terminal extracellular domain of the C5aR (met1-Asn31). Clone P12/1 does not inhibit the binding of C5a to its receptor.
Store at 4C in the dark.
IgG2a
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only