Learn More
Complement C3 Antibody (11H9), DyLight 405, Novus Biologicals™
Rat Monoclonal Antibody
Marca:  Novus Biologicals NB200-540V
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215453
  • 628.49€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
Complement C3 Monoclonal specifically detects Complement C3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
Complement C3
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
C3
This Complement C3 Antibody (11H9) was developed against C57BL/6 thymocytes saturated with rat anti-Thy-1 monoclonal antibody of IgG2b subclass (RmT1).
0.1 mL
Angiogenesis, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Mesenchymal Stem Cell Markers, Neurodegeneration, Neuroscience, Signal Transduction
718
Mouse
Purified
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
11H9
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product
Rat
Protein G purified
RUO
Primary
Mouse Complement C3 and its activation products, C3b, iC3b, C3d and C3dg
Store at 4C in the dark.
IgG2a