Learn More
Nabertherm™ Compact Tube Furnace with B510 Controller
Product Code. 17388089 Shop All Nabertherm Products
Change view
Click to view available options
Diameter (Metric) Process Tube:
100 mm
120 mm
170 mm
Heated Zone:
500 mm
750 mm
1000 mm
Temperature (Metric):
1200°C
1300°C
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Process Tube Heated Zone Temperature (Metric) Unit Size
17388089 100 mm 750 mm 1300°C Each
17318089 120 mm 500 mm 1200°C Each
17328089 120 mm 500 mm 1300°C Each
17378089 170 mm 750 mm 1200°C Each
17338099 170 mm 1000 mm 1200°C Each
17348099 170 mm 1000 mm 1300°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17388089 Supplier Nabertherm™ Supplier No. R472L3AN
 Restricted Item
Fecha estimada de envoi: 12-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
  • Excellent price - performance ratio
  • Can also be equipped with gas supply system 1 (no vacuum operation) and gas supply system 2 (for vacuum operation)
  • Temperature, max. 1200°C or 1300°C
  • Single-zoned design as standard with controller B 510
  • Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
  • Dual shell housing made of sheets of textured stainless steel
  • Insulation made of non-classified fibre material
Please Note: A furnace below 2.9 kW will be delivered with a standard UK plug. Anything above 2.9 kW will be delivered with the appropriate CEE plug. For anything above 2.9 kW, please ensure you have the required socket prior to purchase.
Specifications
Controller Type B510
Heated Zone 750 mm
Temperature (Metric) 1300°C
Type Compact Tube Furnace
Diameter (Metric) Process Tube 100 mm