Learn More
Brady™ Compact Lockout Padlocks
Product Code. 15621127 Shop All Brady Products
Change view
Click to view available options
Color:
Red
Unit Size:
Pack of 6
Product Code. Color Unit Size
15621127 Red Pack of 6
1 options
This item is not returnable. View return policy
Product Code. 15621127 Supplier Brady™ Supplier No. 814116
  • ONLINE EXCLUSIVE PRICE
    123.00€
    107.14€ / paquete de 6
    Ahorro 15.86€
    12% Descuento
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Brady Compact Safety Padlocks are a lightweight range of padlocks suitable for energy isolation programs.
  • They compliment the Brady Safety Padlock and have the same features
  • Unique cylinder is insulated to protect workers from shocks when key is inserted and is 5-pin precision-machined for more unique key cuts and better tamper resistance
  • Reserved, paracentric keyway provides optimal security and the special design prohibits the key from being released until the padlock shackle is closed
  • Locks are compact and lightweight and resist impact, temperature, chemicals and corrosion
  • Their non-conductive, non-sparking body features a ribbed design for easier gripping and handling
  • 1 key supplied with each lock
  • Key retaining (Key won’t release until the padlock shackle is securely closed)
  • Custom keying and charting is available
Specifications
Alternate Part Number 814116
Color Red
Depth (Metric) 16 mm
Height (Metric) 64 mm
Height (Metric) Shackle 25.4 mm
Width (Metric) 32 mm
Material (Body) Reinforced Nylon
Material (Shackle) Aluminium