Learn More
Plas Labs™ Compact Glove Box
Product Code. 15164352
This item is not returnable. View return policy
Product Code. 15164352 Supplier Plas Labs™ Supplier No. 830CP31/EXP
  • ONLINE EXCLUSIVE PRICE
    16594.83€
    10540.95€ / paquete de 1
    Ahorro 6053.88€
    36% Descuento
 Restricted Item
Fecha estimada de envoi: 22-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
size C, ID 1220mm x 740mm x 810mm, includes 1 extra pair of 800-GH gloves and two replacement filters, fitted with Schuko plug CEE 7/7