Learn More
Gilac™ Compact Bread Container
Product Code. 17968671
Change view
Click to view available options
Color:
Beige
White
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17968671 White Each
17908431 Beige Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17968671 Supplier Gilac™ Supplier No. G180621
  • ONLINE EXCLUSIVE PRICE
    140.90€
    92.35€ / paquete de 1
    Ahorro 48.55€
    34% Descuento
Fecha estimada de envoi: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Compact Bread Container is suitable for the new baguette size. Perforated to keep products fresh.
  • Compact bread container, suitable for the new baguette size.
  • Perforated to keep products fresh.
  • Height of two 120 L nested bread containers: 73.5 cm/Stacked: 109.5 cm
Specifications
Product Type Bread Container
Material Polyethylene
Color White
Volume (English) 31.7 gal.
Volume (Metric) 120 L