Learn More
3M™ Comfort Belt SS-626 for 3M™ Powered Air Respirator
Product Code. 17756973 Shop All 3M Products
Change view
Click to view available options
Belt; For Use With: 3M™ PAPR Systems
Product Code.
17756973 Belt; For Use With: 3M™ PAPR Systems
1 options
This item is not returnable. View return policy
Product Code. 17756973 Supplier 3M™ Supplier No. 7100236838
  • ONLINE EXCLUSIVE PRICE
    381.00€
    322.11€ / paquete de 4
    Ahorro 58.89€
    15% Descuento
Fecha estimada de envoi: 30-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
3M™ Comfort Belt SS-626 is our comfort belt designed for use with 3M™ Powered Air Respirator PF-600E.
  • Comfort belt features extra padding for user ease
  • Suitable for use with 3M™ Powered Air Respirator PF-600E
  • Loops around waist and comfortably secures turbo to user
  • Snap buckle for secure fastening
TRUSTED_SUSTAINABILITY
Specifications
Product Type Belt
For Use With (Equipment) Compatible with 3M™ PAPR Systems
Model SS-626