Learn More
Fisherbrand™ Combs for Mini-Plus Wide Format Horizontal Gel Units, 2mm Thick
For use with Mini-Plus Wide Format Horizontal Gel Units.
Marca:  Fisherbrand
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 11513382
  • 30.70€ / cada uno
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • To maximise sample volume
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Especificación
Especificación
2mm
Comb
1 Each
81μL
Blue
Mini-Plus Wide Format Horizontal Gel Units
12
Videos