missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen Type II Mouse Anti-Human, Unlabeled, Clone: II-4C11, MP Biologicals
Mouse Monoclonal Antibody
Marca: MP Biomedicals
Detalles adicionales : Peso : 0.00010kg
Descripción
Collagen Type II Monoclonal antibody specifically detects Collagen Type II in Human samples. It is validated for Immunohistochemistry-frozen, Immunohistochemistry-paraffin, Western Blot.Especificaciones
Collagen Type II | |
Monoclonal | |
1.0mg/mL | |
Mouse | |
500 μg | |
Store at -20°C. Avoid freeze/thaw cycles. | |
IgG1 κ |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
II-4C11 | |
Unconjugated | |
Collagen Type II from human costal cartilage | |
Primary | |
Purified |