Learn More
Coilin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA566995
Detalles adicionales : Peso : 0.01000kg
Descripción
Immunogen sequence: ASETVRLRLQF DYPPPATPHC TAFWLLVDLN RCRVVTDLIS LIRQRFGFSS GAFLGLYLEG GLLPPAESAR LVRDNDCLRV KLEERGVAEN SVVISNGDIN LSLRKAKKRA FQLEEGEETE PDC Highest antigen sequence identity to the following orthologs - mouse 81%, rat 79%.
The protein encoded by this gene is an integral component of Cajal bodies (also called coiled bodies). Cajal bodies are nuclear suborganelles of varying number and composition that are involved in the post-transcriptional modification of small nuclear and small nucleolar RNAs. The N-terminus of the coilin protein directs its self-oligomerization while the C-terminus influences the number of nuclear bodies assembled per cell. Differential methylation and phosphorylation of coilin likely influences its localization among nuclear bodies and the composition and assembly of Cajal bodies. This gene has pseudogenes on chromosome 4 and chromosome 14.Especificaciones
Coilin | |
Polyclonal | |
Unconjugated | |
COIL | |
C79982; CLN80; COIL; Coilin; coilin p80; hypothetical protein LOC507580; LOW QUALITY PROTEIN: coilin; nuclear phosphoprotein of Cajal body; p80; p80-coilin; wu:fb08h11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8161 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P38432 | |
COIL | |
Recombinant Human Coilin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only