missing translation for 'onlineSavingsMsg'
Learn More

Coagulation Factor III/Tissue Factor Antibody (034), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Bio-Techne NBP2-90576G

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215498

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Coagulation Factor III/Tissue Factor Monoclonal specifically detects Coagulation Factor III/Tissue Factor in Mouse samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

Coagulation Factor III/Tissue Factor
Monoclonal
DyLight 488
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Mouse
Purified
Western Blot, ELISA
34
Western Blot, ELISA
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Coagulation Factor III/Tissue Factor/ F3 (NP_034301.3; Met 1-Glu 251).
0.1 mL
Cancer
2152
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales