Learn More
Novus Biologicals™ CNPY2 Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-09323
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215662
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
The CNPY2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC29441 and based on accession number NM_14255. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens canopy 2 homolog (zebrafish) (CNPY2), mRNA.
Native