missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNKSR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10885-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein containing several motifs involved in protein-protein interaction, including PDZ, PH (Pleckstrin homology), and SAM (sterile alpha motif) domains. The encoded protein acts as a scaffold component for receptor tyrosine kinase signaling and may mediate crosstalk between different signaling pathways. Alternative splicing results in multiple transcript variants.Especificaciones
CNKSR1 | |
Polyclonal | |
Unconjugated | |
CNKSR1 | |
CNK, CNK homolog protein 1, CNK1, CNKSR1, Connector enhancer of KSR 1, Connector enhancer of KSR like, hCNK1, KSR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10256 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q969H4 | |
CNKSR1 | |
CNKSR1 Fusion Protein Ag1333 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |