Learn More
Cytiva CNBr-Activated Sepharose™ Medium
Product Code. 11514905 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
15 g
Unit Size:
15g
Product Code. Quantity Unit Size
11514905 15 g 15g
1 options
This item is not returnable. View return policy
Product Code. 11514905 Supplier Cytiva Supplier No. 17043001
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Use for coupling antibodies or other large proteins containing -NH2 groups to the Sepharose media. CNBr-Activated Sepharose™ Medium is a pre-activated media perfect for custom affinity applications.
  • Allows spontaneous and rapid coupling of proteins without the need for an intermediate spacer arm
  • Multipoint attachment resulting in a chemically stable product
TRUSTED_SUSTAINABILITY
Specifications
Ligand Coupling Method Large Protein
Matrix 4% Agarose
Particle Size 90 μm
pH 2 to 11
Quantity 15 g
Content And Storage 2°C to 8°C
For Use With (Application) For coupling ligands containing primary amino groups