missing translation for 'onlineSavingsMsg'
Learn More

CMTM8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 15039-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16814014

  • 319.00€ / 150 microlitros

Envío estimado: 20-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown.
Especificaciones

Especificaciones

CMTM8
0.53 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IZV2, Q9CZR4
CKLFSF8, CKLFSF8 V2, CMTM8
Rabbit
IgG
150 μL
-20°C
Primary
152189, 301045, 70031
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
CMTM8
Liquid
Cmtm8
CMTM8 Fusion Protein Ag6776
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados