Learn More
Fisherbrand™ Clinical Bundle
Product Code. 30130309 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Capacity:
8 x 3 to 15 mL
Unit Size:
1 set
Product Code. Capacity Unit Size
30130309 8 x 3 to 15 mL 1 set
1 options
This item is not returnable. View return policy
Product Code. 30130309
  • ONLINE EXCLUSIVE PRICE
    1105.00€
    827.84€ / 1 sets
    Ahorro 277.16€
    25% Descuento
View all promotions
 Restricted Item
Fecha estimada de envoi: 18-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Optimize your lab efficiency with this comprehensive clinical bundle.
Spin samples effortlessly with a high-performance centrifuge, keep your tubes organized and accessible with a multi-tube rack, and safeguard your workspace with a durable lab mat. This bundle provides everything you need for streamlined and precise sample handling. This bundle includes the following:
Fisherbrand™ Clinical Centrifuge - #17801727
  • This compact, quiet clinical benchtop centrifuge offers adjustable speeds (500-6,000 RPM), programmable memory, and built-in safety features, maximizing functionality and workspace efficiency.
Fisherbrand™ Clinical 4-Way Tube Rack, Assorted - #16340402
  • This rack holds 13 and 17 mm tubes securely, is made of durable, chemical-resistant polypropylene, and withstands water baths and freezers. Customizable and adaptable for any setup.
Fisherbrand™ Clinical Lab Mat - #16361648
  • This lab mat safely facilitates workflow processes with organization and easy clean up, while keeping benchtops clean and safe from stains or spills.
Specifications
Includes Tube Spacers, Adapter Sleeves, 8-Place Rotor, Power Adapter
Capacity 8 x 3 to 15 mL
Max. RCF 4,668 x g
Max. Speed 6,000 rpm
Refrigerated No
Certifications/Compliance CE, UKCA, RoHS3, WEEE
Timer 30 sec. to 60 min., or Continuous
Wattage 93 W
Weight (English) 9.93 lb.
Weight (Metric) 4.5 kg
Voltage 100/240 V
Product Type Clinical Centrifuge
Temperature Range (Metric) 2°C to 40°C