Learn More
Invitrogen™ Click-iT™ sDIBO Alkyne for Antibody Labeling
The Click-iT Biotin sDIBO Alkyne for Antibody Labeling is optimized for easy attachment to azido modified antibodies using copper-free Click chemistry.
Marca:  Invitrogen™ C20032
Detalles adicionales : Peso : 0.01900kg
 Ver más versiones de este producto
Código de producto. 15920583
  • 149.00€ / cada uno
Fecha de envío estimada
del proveedor 02-04-2019
Añadir a la cesta
Descripción
Descripción
The Click-iT sDIBO Alkyne for Antibody Labeling is optimized for easy attachment to azido modified antibodies using copper-free Click chemistry. This sDIBO label can be used with antibodies that have been modified using the SiteClick Antibody Azido Modification Kit or antibodies that have been engineered to contain azido moieties. These sDIBO alkynes are improved versions of our original DIBO cyclooctynes, yielding conjugates that are less 'sticky' and give lower signal background in biological samples.
This modular labeling system gives you the option to choose different fluorescent labels for your antibody and attach another molecule via streptavadin or your own molecule via amine-reactive or amine-containing moieties depending on your assay.
Custom SiteClick Antibody Labeling Service and sDIBO labels
If you have an antibody that is considered 'difficult to label' or has lost activity after labeling using a conventional method, please contact our custom service representatives to determine whether the SiteClick Antibody Labeling Service would be right for your antibody. We offer complete custom SiteClick antibody labeling services with the option of multiple detection molecules including biotin, Alexa Fluor dyes, Qdot fluorophores, R-PE, chelates for PET imaging, and many others.
Order Info
Store in freezer (-5 to -30°C) and protect from light.
Especificación
Especificación
Click-iT™ SDP Ester sDIBO Alkyne for Antibody Labeling
Antibody Labeling
1 Kit
Kit
Store in freezer (-5 to -30°C) and protect from light.
Videos