missing translation for 'onlineSavingsMsg'
Learn More

CLIC4 Mouse anti-Human, Porcine, Rat, Clone: 1B7F9, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66343-1-IG-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818135

  • 109.00€ / 20 microlitros

Envío estimado: 22-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintece of intracellular pH, and regulation of cell volume. Chloride intracellular channel 4 (CLIC4) protein, encoded by the CLIC4 gene, is a member of the p64 family; the gene is expressed in many tissues and exhibits a intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells).
Especificaciones

Especificaciones

CLIC4
1B7F9
Unconjugated
CLIC4
Liquid
CLIC4
CLIC4 Fusion Protein Ag2942
Protein G
RUO
Antibody
Monoclonal
Porcine, Rat, Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y696, Q9Z0W7
CLIC4, CLIC4L, H1, huH1, MTCLIC, p64H1
Mouse
IgG1
20 μL
-20°C
Primary
100622787, 25932, 83718
Videos
SDS
Documentos
Certificados