Learn More
CLEC4G Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18173-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a glycan-binding receptor and member of the C-type lectin family which plays a role in the T-cell immune response. Multiple transcript variants encoding different isoforms have been found for this gene.Especificaciones
CLEC4G | |
Polyclonal | |
Unconjugated | |
CLEC4G | |
CLEC4G, LP2698, LSECtin, UNQ431 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
339390 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UXB4 | |
CLEC4G | |
CLEC4G Fusion Protein Ag12837 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |