missing translation for 'onlineSavingsMsg'
Learn More

CLEC3B/Tetranectin Antibody (006), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90147AF350

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213406

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

CLEC3B/Tetranectin Monoclonal specifically detects CLEC3B/Tetranectin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

CLEC3B/Tetranectin
Monoclonal
Alexa Fluor 350
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
No cross-reactivity in ELISA with: Mouse CLEC14A
Store at 4°C in the dark.
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
6
Immunohistochemistry, Immunohistochemistry-Paraffin
C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin
This antibody was obtained from a rabbit immunized with purified, recombinant Human CLEC3B/Tetranectin (AAX37102.1; Met1-Val202).
0.1 mL
Cancer
7123
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales