missing translation for 'onlineSavingsMsg'
Learn More

CLEC16A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 26257-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16814775

  • 319.00€ / 150 microlitros

Envío estimado: 21-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the C-type lectin domain containing family. Single nucleotide polymorphisms in introns of this gene have been associated with diabetes mellitus, multiple sclerosis and rheumatoid arthritis. Multiple transcript variants encoding different isoforms have been found for this gene.
Especificaciones

Especificaciones

CLEC16A
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q2KHT3, Q80U30
CLEC16A, Gop 1, KIAA0350, Protein CLEC16A
Rabbit
IgG
150 μL
-20°C
Primary
23274, 74374
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CLEC16A
Liquid
Clec16a
CLEC16A Fusion Protein Ag24268
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documentos
Certificados