Learn More
Brannan™ Classroom Wall Kit - Thermometer and Chart with 3 Point Traceable Calibration Certificate
Product Code. 17842988
Change view
Click to view available options
Temperature (Metric) Operating:
-20°C to +50°C
Unit Size:
Each
Product Code. Temperature (Metric) Operating Unit Size
17842988 -20°C to +50°C Each
1 options
This item is not returnable. View return policy
Product Code. 17842988 Supplier Brannan™ Supplier No. 25/702/0 + 49/101/0
  • ONLINE EXCLUSIVE PRICE
    126.00€
    116.71€ / paquete de 1
    Ahorro 9.29€
    7% Descuento
Fecha estimada de envoi: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Single packed classroom wall thermometer kit with laminated chart. Complete with 3 point traceable calibration.
  • Colorful 380 mm plastic wall thermometer with chart for recording temperatures
  • Brightly designed, ideal for education
  • Supplied with calibration certificate
  • Laminated chart making in easy to clean for multiple uses
  • Made in UK
Specifications
Certifications/Compliance UK National Standards ISO 9001
For Use With (Application) Wall Thermometer with chart
Temperature (Metric) Operating -20°C to +50°C
Product Type Wall Thermometer Kit