missing translation for 'onlineSavingsMsg'
Learn More

CKM Mouse anti-Human, Mouse, Rat, Clone: 2G3F6, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60177-1-IG-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817155

  • 399.00€ / 150 microlitros
Fecha estimada de envío: 17-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Creatine kinase isoenzymes play a central role in energy transduction in tissues with large, fluctuating energy demands, such as skeletal muscle, heart, brain and spermatozoa. The CKM isoform, predomit in skeletal muscle and heart tissue, is a cytoplasmic enzyme involved in energy homeostasis and is an important serum marker for myocardial infarction. CKM reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in striated muscle as well as in other tissues, and as a heterodimer with a similar brain isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family.
Especificaciones

Especificaciones

CKM
Monoclonal
2.03 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P00564, P06732, P07310
Ckm
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG2a
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2G3F6
Unconjugated
CKM
CKMM, Creatine Kinase MM, Creatine kinase M chain, Creatine kinase M type, creatine kinase, muscle, M CK
Mouse
Protein A
RUO
1158, 12715, 24265
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales