Learn More
Ácido cis-3-(Boc-amino)ciclohexanocarboxílico, 97 %, Thermo Scientific Chemicals
Catalog Number p-7041485
Click to view available options
Cantidad:
250 mg
1 g
222530-33-8
C12H21NO4
243.303
MFCD01862295
JSGHMGKJNZTKGF-BDAKNGLRSA-N
1r,3s-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 1r,3s-3-boc-amino-cyclohexanecarboxylic acid, cis-+/--3-boc-amino cyclohexanecarboxylic acid, boc-cis-3-aminocyclohexanecarboxylic acid, cis-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3s-rel-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3s-3-boc-amino cyclohexanecarboxylic acid, cis-3-boc-amino cyclohexanecarboxylic acid, cis-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid
16218560
Ácido (1R,3S)-3-[(2-metilpropan-2-il)oxicarbonilamino]ciclohexano-1-carboxílico
CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O
222530-33-8
C12H21NO4
243.303
MFCD01862295
JSGHMGKJNZTKGF-BDAKNGLRSA-N
1r,3s-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 1r,3s-3-boc-amino-cyclohexanecarboxylic acid, cis-+/--3-boc-amino cyclohexanecarboxylic acid, boc-cis-3-aminocyclohexanecarboxylic acid, cis-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3s-rel-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3s-3-boc-amino cyclohexanecarboxylic acid, cis-3-boc-amino cyclohexanecarboxylic acid, cis-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid
16218560
Ácido (1R,3S)-3-[(2-metilpropan-2-il)oxicarbonilamino]ciclohexano-1-carboxílico
CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material cis-3-(Boc-amino)cyclohexanecarboxylic acid
Punto de fusión 143°C to 147°C
Formula Weight (peso de la fórmula) 243.29
Porcentaje de pureza 97%