Learn More
Steris™ CIP 220
Product Code. 15103726
Change view
Click to view available options
Volume (Metric):
18.93 L
Unit Size:
18.9 L
Product Code. Volume (Metric) Unit Size
15103726 18.93 L 18.9 L
1 options
This item is not returnable. View return policy
Product Code. 15103726 Supplier Steris™ Supplier No. 1D2205
 Restricted Item
Fecha estimada de envoi: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Product Type Acid-based Cleanser
Volume (Metric) 18.93 L
compliance-icons compliance-icons
Product Identifier
  • CIP 220
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
  • MIXTURE LIST-Contain: Hydroxyacetic acid