Learn More
Proteintech ChromoTek pSpot1 Vector
Product Code. 17333323
Change view
Click to view available options
Quantity:
1.25 μg
Unit Size:
1.25µg
Product Code. Quantity Unit Size
17333323 1.25 μg 1.25µg
1 options
This item is not returnable. View return policy
Product Code. 17333323 Supplier Proteintech Supplier No. EV1
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
pSpot1 vector, E.coli, Spot N-term, Kan., high expression.
  • The plasmid pSpot1 is an expression vector for Spot-Tag™ fusion proteins in E. coli. After cloning the protein of interest (POI) into the multiple cloning site (MCS) provided by pSpot1, the Spot-Tag™ (sequence: PDRVRAVSHWSS) will be fused to the N-terminus of the POI.
Specifications
Quantity 1.25 μg