Learn More
Macherey-Nagel™ Chrombond™ RS 40 C18 ec Polypropylene Flash Column
Product Code. 10339384 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
171 mm
Unit Size:
Each
Product Code. Length (Metric) Unit Size
10339384 171 mm Each
1 options
This item is not returnable. View return policy
Product Code. 10339384 Supplier Macherey-Nagel™ Supplier No. 732813
  • ONLINE EXCLUSIVE PRICE
    173.00€
    163.80€ / paquete de 1
    Ahorro 9.20€
    5% Descuento
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Length (Metric) 171 mm
Material Polypropylene
Type Flash Column
Column Type Flash Column
For Use With (Equipment) Directly Used in The Teledyne Isco Companion, Rf