Learn More
Macherey-Nagel™ Chromafix™ C18 Cartridge
Product Code. 10755952 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
240 mg
480 mg
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Unit Size
10755952 240 mg Pack of 50
10628621 480 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10755952 Supplier Macherey-Nagel™ Supplier No. 731801
  • ONLINE EXCLUSIVE PRICE
    197.62€
    178.50€ / paquete de 50
    Ahorro 19.12€
    9% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Base Material Silica
Carbon Load 14%
Bed Weight 240 mg
For Use With (Application) Non-Polar Compounds, Pesticides
Particle Size 45 μm
Sorbent C18
Type Chromafix C18 Cartridge
Pore Size 60 Å
Quantity 50 Pack
Surface Area (Metric) 500 m2/g