Learn More
Macherey-Nagel™ Chromabond™ Polyethylene Frit
Product Code. 12817921 Shop All Macherey-Nagel Products
Change view
Click to view available options
Volume (Metric):
1 mL
3 mL
6 mL
15 mL
30 mL
45 mL
70 mL
150 mL
Unit Size:
Pack of 250
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12817921 150 mL Pack of 250
10766142 15 mL Pack of 250
10338311 1 mL Pack of 250
10072871 30 mL Pack of 250
10639581 3 mL Pack of 250
10093151 45 mL Pack of 250
10154801 6 mL Pack of 250
10418981 70 mL Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 12817921 Supplier Macherey-Nagel™ Supplier No. 730475
Fecha estimada de envoi: 19-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Assemble SPE (solid-phase extraction) columns selecting from a range of accessories. Chromabond™ Polyethylene Frits for polyethelyne columns come in a varitey of sizes.
Accessory for SPE (solid-phase extraction)
For individual packing of SPE columns with Chromabond™ adsorbents
Specifications
For Use With (Equipment) Polypropylene Columns
Type Frit
Volume (Metric) 150 mL
Material Polyethylene
Quantity 250 Pack