missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHPF C-terminal Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
121.38€ - 325.38€
Especificaciones
Antígeno | CHPF C-terminal |
---|---|
Concentración | 0.9 mg/mL |
Aplicaciones | Western Blot, Immunohistochemistry (Paraffin) |
Clasificación | Polyclonal |
Conjugado | Unconjugated |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16818663
|
Proteintech
11524-1-AP-20UL |
20 μL |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
16808663
|
Proteintech
11524-1-AP-150UL |
150 μL |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
Descripción
Chondroitin sulfate synthases (CHSYs) synthesize chondroitin sulfate, a glycosaminoglycan expressed on the surface of most cells and in extracellular matrices. Glycosaminoglycan chains are covalently linked to various of core protein families and regulate many biologic processes, including extracellular matrix deposition, cell proliferation and recognition, and morphogenesis. The CHSY family includes CHSY1, CHSY2 and CHSY3. CHSY1 and CHSY3 display both glucuronyltransferase and N-acetylgalactosaminyltransferase activities, while CHSY2 is required for chondroitin polymerizing activity. CHSY2 localizes to the Golgi apparatus and is expressed ubiquitously, with highest expression observed in pancreas, ovary, brain, heart, skeletal muscle, colon, kidney, liver, stomach, small intestine and placenta.Especificaciones
CHPF C-terminal | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q6IQX7, Q8IZ52 | |
316533, 74241, 79586 | |
CHPF C-terminal Fusion Protein Ag2088 | |
Primary | |
-20°C | |
CHPF |
0.9 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Chondroitin sulfate synthase 2, CHPF, CHSY2, CSS2 | |
Chpf | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |